Learn More
Poulten Graf™ Fortuna™ Optifix™ Basic Dispenser
Description
For dosing of the most common chemicals, acids and crytallizing liquids (not suitable for solvents). Poulten Graf™ PTFE/PP/Fiberglass/Teflon/Ceramic Fortuna™ Optifix™ Basic Dispensers have a dispensing cylinder and piston made of glass.
- Manufactured according to DIN EN ISO 9001:2008 using high quality materials
- With solid PTFE encased dispensing piston made of glass
- Cylinder coated with plastic to protect against breakage
- With certificate, individually numbered
- Ergonomic finger protection cap made of PP with volume setting system
- Graduation on the scale is lasered and easy to read
- Fast and easy volume setting with additional fine adjustment
Specifications
Specifications
| Product Type | Fortuna Optifix Basic Dispenser |
| Color | Clear |
| Material | PTFE, Polypropylene, Fiberglass, Teflon, Ceramic |
| Volume (Metric) | 60 to 300 mL |
| For Use With (Application) | For dispensing aqueous solutions, acids and crystallizing liquids |
| Quantity | Each |