Description
- PTFE construction
- Polypropylene valve
- With integrated hose nozzle
Specifications
Specifications
| Plug Bore | 5 mm |
| Material | PTFE |
| Fits Tubing ID | 9 mm |
| Includes | Integrated hose nozzle |
| Plug Material | Polypropylene |