Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Product Code. 16583592
Change view
Click to view available options
Length (Metric):
5 mm
8 mm
10 mm
12 mm
15 mm
20 mm
25 mm
30 mm
40 mm
60 mm
70 mm
80 mm
Diameter (Metric):
2 mm
3 mm
4.5 mm
6 mm
7 mm
8 mm
10 mm
Unit Size:
Pack of 10
Product Code. Length (Metric) Diameter (Metric) unitSize
16583592 5 mm 2 mm Pack of 10
16513602 10 mm 3 mm Pack of 10
10264171 10 mm 6 mm Pack of 10
10375661 12 mm 4.5 mm Pack of 10
10674602 15 mm 6 mm Pack of 10
10375711 25 mm 6 mm Pack of 10
10789421 30 mm 6 mm Pack of 10
10204271 40 mm 8 mm Pack of 10
10224221 20 mm 7 mm Pack of 10
10480343 60 mm 7 mm Pack of 10
16533602 70 mm 10 mm Pack of 10
16543602 80 mm 10 mm Pack of 10
16503602 8 mm 3 mm Pack of 10
13 options
Product Code. 16583592
Brand: VITLAB™ 300497
Only null left
Add to basket
Add to basket
Ensure long-lasting durability and prevent demagnetization of Vitlab™ PFTE Polygonal Magnetic Stirring Bars via proper storage and not dropping them on hard surfaces (e.g. steel).
  • Permanent magnet AlNiCo V core
  • Angled shape gives rise to significant turbulence
  • Achieves effective mixing, even at low spin rates
  • FTE
Specifications
For Use With (Application) For preparation, separation and concentration
Includes Permanent magnet AlNiCo V core
Coating PTFE
Diameter (Metric) 2 mm
Length (Metric) 5 mm
Material PTFE
Quantity 5/Pk.
Shape Polygon
Type Magnetic Stir Bar