Learn More
VITLAB™ PTFE Polygonal Magnetic Stirring Bar
Description
- Permanent magnet AlNiCo V core
- Angled shape gives rise to significant turbulence
- Achieves effective mixing, even at low spin rates
- FTE
Specifications
Specifications
| For Use With (Application) | For preparation, separation and concentration |
| Includes | Permanent magnet AlNiCo V core |
| Diameter (Metric) | 8 mm |
| Length (Metric) | 40 mm |
| Material | PTFE |
| Quantity | 10/Pk. |
| Shape | Polygon |