Learn More
Advantec PTFE O-ring
Product Code. 14891211 Shop All Cole-Parmer Products
Change view
Click to view available options
Material:
PTFE
Unit Size:
Each
Product Code. Material unitSize
14891211 PTFE Each
1 options
Product Code. 14891211
Brand: Advantec 0293015
This item requires a price update prior to being ordered.
Request a Price for this item here.
Optimize filtration with this PTFE O-ring.
  • Choose from a variety of options.
  • Find items compatible with filter holders and stirred cells.
Specifications
For Use With (Equipment) SS Filter Holders, KS13
Quantity 1 Ea.
Product Type O-Ring
Material PTFE
Max. Pressure 1000 psi