Learn More
SVL™ Rotulex™ PTFE O-ring for Spherical Joint
Product Code. 12099022
Click to view available options
Bore Size:
5mm
9 mm
15 mm
20 mm
25 mm
40 mm
Unit Size:
Each
Product Code. 12099022
Brand: SVL RO41
Only null left
Add to basket
Add to basket
Offers a 'grease-free' spherical joint. Rotulex™ PTFE O-ring for Spherical Joint is highly resistant to chemical corrosion. Can be used at temperatures up to 200 °C. PTFE covered O-ring fits into a female cup joint which can be retained together with spring loaded metal clip.
  • Offers a 'grease-free' spherical joint
  • PTFE pathway is highly resistant to chemical corrosion
  • Can be used at temperatures up to 200°C
  • PTFE covered O-ring fits into a female cup joint
  • Joint can be retained together with spring loaded metal clip
TRUSTED_SUSTAINABILITY
Specifications
Bore Size 25 mm
For Use With (Equipment) RM:RF41 Glass Joint
Quantity 1 Each
Diameter (Metric) Outside 28 mm (Shank), 41 mm (Spherical)
Material PTFE