Learn More
Bohlender™ BOLA™ PTFE Moon Shaped Stirrer Shafts
Product Code. p-8005476
Change view
Click to view available options
Length (Metric):
350 mm
450 mm
510 mm
600 mm
800 mm
Diameter (Metric) Shaft:
8 mm
10 mm
16 mm
Standard Taper Joint:
24/29
29/32
45/40
Unit Size:
Each
Product Code. Length (Metric) Diameter (Metric) Shaft Standard Taper Joint unitSize
10751421 350 mm 10 mm 29/32 Each
10761421 450 mm 10 mm 29/32 Each
10707692 510 mm 10 mm 29/32 Each
10679211 800 mm 16 mm 45/40 Each
10518462 350 mm 8 mm 24/29 Each
10696491 600 mm 10 mm 29/32 Each
10635921 450 mm 8 mm 29/32 Each
10053910 600 mm 8 mm 29/32 Each
8 options
Product Code. 10751421
Brand: Bohlender™ C37612
Only null left
Add to basket
Add to basket
Use these tilting half-moon blades for stirring in round bottom flasks with ground joint necks. Bohlender™ BOLA™ PTFE Moon Shaped Stirrer Shafts offer tangential flow with little turbulence and universal chemical resistance.
  • PTFE-jacketed stainless steel shaft with tilting half-moon stirrer blade
  • Double-sided groove and access for the stirrer shaft completely made of PTFE
  • Temperature resistance: -200 to +250°C
  • Blades are available separately and can be mounted additionally
Specifications
Diameter (Metric) Shaft 10 mm
For Use With (Application) The tilting half-moon blade is ideal for stirring in round-bottom flasks with ground joint necks.
Length (Metric) 350 mm
Quantity 1 Each
Standard Taper Joint 29/32
Type Moon Shaped Stirrer Shaft