Learn More
Bohlender™ BOLA™ PTFE Moon Shaped Stirrer Shafts
Click to view available options
Length (Metric):
350 mm
450 mm
510 mm
600 mm
800 mm
Diameter (Metric) Shaft:
8 mm
10 mm
16 mm
Standard Taper Joint:
24/29
29/32
45/40
Unit Size:
Each
Description
- PTFE-jacketed stainless steel shaft with tilting half-moon stirrer blade
- Double-sided groove and access for the stirrer shaft completely made of PTFE
- Temperature resistance: -200 to +250°C
- Blades are available separately and can be mounted additionally
Specifications
Specifications
| Diameter (Metric) Shaft | 10 mm |
| For Use With (Application) | The tilting half-moon blade is ideal for stirring in round-bottom flasks with ground joint necks. |
| Length (Metric) | 510 mm |
| Quantity | 1 Each |
| Standard Taper Joint | 29/32 |
| Type | Moon Shaped Stirrer Shaft |