Learn More
Fisherbrand™ PTFE Micro Magnetic Stir Bar
Product Code. 10180301 Shop All Fisherbrand Products
Click to view available options
Length (Metric):
2 mm
3 mm
5 mm
6 mm
7 mm
8 mm
10 mm
13 mm
15 mm
20 mm
30 mm
Diameter (Metric):
1.5 mm
2 mm
3 mm
Unit Size:
Each
Pack of 10
Product Code. 10180301
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Designed for the smallest containers. Fisherbrand™ PTFE Micro Magnetic Stir Bar are autoclavable.
  • Supplied in reusable snap cap vials.
  • For stirring in test tubes, small size beakers, or other containers.
  • Autoclavable.
Specifications
Autoclavable Autoclavable
Certifications/Compliance Certificate of Conformance, Certificate of Analysis, Product Data Sheet, Material Safety Data Sheet (MSDS)
Color White
Description PTFE encapsulated Alnico V magnet core
For Use With (Equipment) Containers & Vessels / Stirrer Hot Plates
Coating PTFE
Diameter (Metric) 2 mm
Length (Metric) 5 mm
Quantity 10/Pk.
Shape Cylinder
Type PTFE Stirrer Bars Micro