Learn More
Cole-Parmer™ PTFE Manifolds
Product Code. p-8007918
Change view
Click to view available options
Length (Metric):
120.65 mm
186.4 mm
199.3 mm
238.3 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
11785268 199.3 mm Each
11715278 238.3 mm Each
11745268 120.65 mm Each
11705278 186.4 mm Each
4 options
Product Code. 11785268
Brand: Cole-Parmer™ 3152121
Specifications
Type Manifold
Inlet 3/8 in.
Material PTFE
No. of Ports 8
Port Size (Metric) 5.08 mm
Length (Metric) 199.3 mm
Width (Metric) 28.9 mm
Height (Metric) 28.95 mm