Learn More
Cole-Parmer™ PTFE Manifolds
Click to view available options
Length (Metric):
120.65 mm
186.4 mm
199.3 mm
238.3 mm
Unit Size:
Each
Specifications
Specifications
| Type | Manifold |
| Inlet | 1/4 in. |
| Material | PTFE |
| No. of Ports | 6 |
| Port Size (Metric) | 4.3 mm |
| Length (Metric) | 120.65 mm |
| Width (Metric) | 25.4 mm |
| Height (Metric) | 25.4 mm |