Learn More
Cole-Parmer™ PTFE Manifold, 2 outlets
Product Code. p-7138705
Click to view available options
Overall Length (English):
1.75 in.
2 3/8 in.
2.75 in.
Unit Size:
Each
Product Code. 11785258
Brand: Cole-Parmer™ 3152101
Only null left
Add to basket
Add to basket
Organize your fluid paths with these thermoplastic manifolds
  • PTFE Manifold.
  • Chemically-inert PTFE handles the most aggressive chemical solutions.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Material PTFE
Overall Length (English) 1.75 in.
Overall Length (Metric) 4.45 cm
Min. Operating Temperature (English) -200°F
Min. Operating Temperature (Metric) -129°C
Inlet 1/4 in. NPT(F)
Outlet Port 1/8 in. NPT(F)
Width (Metric) 2.54 cm
Width (English) 1 in.
Height (Metric) 2.54 cm
Height (English) 1 in.
Maximum Pressure 120 psi
Max. Operating Temperature (English) 300°F
Max. Operating Temperature (Metric) 150°C
Product Type PTFE Manifold (2 Outlets)