Learn More
Cole-Parmer™ PTFE Manifold
Product Code. 11795268
Change view
Click to view available options
Packaging Quantity:
1/Pk
1/Pk.
Unit Size:
Each
Product Code. Packaging Quantity unitSize
11795268 1/Pk Each
11725278 1/Pk. Each
2 options
Product Code. 11795268
Brand: Cole-Parmer™ 3152123
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Organize your fluid paths with these thermoplastic manifolds
  • Chemically-inert PTFE handles the most aggressive chemical solutions.
  • Available in a variety of configurations to suit a wide range of applications.
Specifications
Material PTFE
Overall Length (Metric) 22.23 cm
Min. Operating Temperature (English) -200°F
Min. Operating Temperature (Metric) -129°C
Packaging Quantity 1/Pk
Tubing Size 1/2 in.
Inlet 1/2 in.
Width (Metric) 3.81 cm
Height (Metric) 3.81 cm
Maximum Pressure 120 psi
Max. Operating Temperature (English) 300°F
Max. Operating Temperature (Metric) 150°C
Product Type PTFE Manifold; 8 Outlets; 1/2 in. Inlet NPT(F)