Learn More
BRAND™ PTFE Magnetic Stirring Bars Retriever
Product Code. 10578401 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
150 mm
250 mm
350 mm
Unit Size:
Each
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) unitSize
10578401 250 mm Each
10759011 350 mm Each
10018310 150 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 10578401 Supplier BRAND™ Supplier No. 137710
Estimated Shipment: 01-09-2026
Add to basket
Add to basket
Manufactured from PTFE. BRAND™ PTFE Magnetic Stirring Bars Retriever has a magnetic core that is fully sealed to prevent leeching.
  • Manufactured from PTFE.
  • Magnetic core is fully sealed to prevent leeching.
  • Manufactured from PTFE.
  • Magnetic core is fully sealed to prevent leeching.
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) 8 mm
Length (Metric) 250 mm
Material PTFE
Quantity 1