Learn More
DWK Life Sciences PTFE Key with Thread, with three part retaining device
Product Code. 15951561 Shop All DWK Life Sciences Products
Change view
Click to view available options
Closure Size:
12.5 NS
14.5 NS
18.8 NS
21.5 NS
24 NS
29.2 NS
Plug Bore:
2 mm
2.5 mm
4 mm
6 mm
8 mm
10 mm
Unit Size:
Each
Pack of 10
Pack of 25
Pack of 50
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Closure Size Plug Bore Unit Size
15951561 21.5 NS 6 mm Pack of 10
15931561 12.5 NS 2 mm Pack of 50
15864618 12.5 NS 2.5 mm Pack of 50
12858041 14.5 NS 2.5 mm Pack of 50
10448572 14.5 NS 4 mm Each
15884618 24 NS 8 mm Pack of 10
12868041 18.8 NS 4 mm Pack of 25
15874618 18.8 NS 6 mm Pack of 25
15894618 29.2 NS 10 mm Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Product Code. 15951561 Supplier DWK Life Sciences Supplier No. 286221206
  • ONLINE EXCLUSIVE PRICE
    2185.00 NOK
    1684.41 NOK / Pack of 10
    Save 500.59 NOK
    22% Off
Estimated Shipment: 14-12-2026
For use with polished DURAN™ disposable taps.
Disposable PTFE valve plugs in combination with the polished DURAN™ disposable valve represent a grease-free alternative. The 3-part tap safety device is already included in the scope of delivery.
  • Made of PTFE with thread and 3-part valve locking device
  • For use with polished DURAN™ one-way taps
  • Grease-free working
Specifications
Plug Bore 6 mm
Material PTFE
Autoclavable Not Autoclavable
Closure Size 21.5 NS
No. per Pack 10/Pk.
For Use With (Application) For use with polished DURAN™ one-way taps