Learn More
Horst™ PTFE HST Model Humidity-Proof Heating Cables
Click to view available options
Length (Metric) Power Cord:
12m
6m
9m
Unit Size:
Each
Specifications
Specifications
| Dimensions (L x W) | 6 Lm |
| Dimensions (L x W x H) | 6 Lm |
| Electrical Requirements | ∼230V Voltage |
| Model | HST |
| Length (Metric) Power Cord | 6m |
| Product Type | Humidity-proof Heating Cable |