Learn More
Horst™ PTFE HBSI Model Hard-Wearing Heating Tape
Specifications
Specifications
| Dimensions (L x W) | 3 Lm |
| Dimensions (L x W x H) | 3 Lm |
| Length (Metric) | 3 m |
| Electrical Requirements | ∼230V Voltage |
| Model | HBSI |
| Product Type | Hard-wearing Heating Tape |