Learn More
Masterflex™ PTFE Gaskets Fitting
Specifications
Specifications
| For Use With (Application) | For pharmaceutical, high-purity water, injectable, food, and beverage applications |
| Quantity | 10 Pack |
| Size (Metric) | 19.05 mm Tri-Clamp |
| Material | PTFE |
| Type | Gasket Fittings |