Learn More
Masterflex™ PTFE Gaskets Fitting
Product Code. 11939426 Shop All Masterflex Products
Change view
Click to view available options
Size (Metric):
19.05 mm Tri-Clamp
25.4 mm Tri-Clamp
Unit Size:
Pack of 10
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Size (Metric) Unit Size
11939426 25.4 mm Tri-Clamp Pack of 10
11959628 19.05 mm Tri-Clamp Pack of 10
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11939426 Supplier Masterflex™ Supplier No. 3054884
  • ONLINE EXCLUSIVE PRICE
    487.00 NOK
    433.28 NOK / Pack of 10
    Save 53.72 NOK
    11% Off
Estimated Shipment: 15-12-2026
Specifications
For Use With (Application) For pharmaceutical, high-purity water, injectable, food, and beverage applications
Quantity 10 Pack
Size (Metric) 25.4 mm Tri-Clamp
Material PTFE
Type Gasket Fittings