Learn More
Bohlender™ PTFE Cylindrical Magnetic Stirring Bar
Product Code. 10637952
Click to view available options
Length (Metric):
6 mm
10 mm
35 mm
Unit Size:
Pack of 10
Pack of 20
Product Code. 10637952
Brand: Bohlender™ C35023
Only null left
Specifications
Certifications/Compliance FDA Certified
For Use With (Application) Cylindrical magnetic stirring bars offer excellent centering and smooth running characteristics
Includes Alnico 5 magnetic core
Coating PTFE
Diameter (Metric) 6 mm
Length (Metric) 35 mm
Material PTFE
Quantity 1/Pk.
Shape Cylinder
Type Magnetic Stir Bar