Learn More
Bochem™ PTFE-Coated Micro Double Spatulas
Product Code. 10306341
Change view
Click to view available options
Length (Metric):
150 mm
185 mm
210 mm
Length (Metric) Blade:
40 mm
50 mm
55 mm
Diameter (Metric) Outer:
1.5 mm
2 mm
2mm
Width (Metric):
3 mm
4 mm
5 mm
6 mm
Unit Size:
Each
Product Code. Length (Metric) Length (Metric) Blade Diameter (Metric) Outer Width (Metric) unitSize
10306341 185 mm 50 mm 2 mm 4 mm Each
10505801 185 mm 50 mm 2mm 5 mm Each
10458442 210 mm 55 mm 2mm 6 mm Each
10202111 150 mm 40 mm 1.5 mm 3 mm Each
4 options
Product Code. 10306341
Brand: Bochem™ 3711
Only null left
Add to basket
Add to basket
Well-suited for small quantity applications and narrow-format tubes, these double-ended, PTFE-coated spatulas reduce material sticking. Bochem™ PTFE-Coated Micro Double Spatulas are also gentle on delicate, scratchable surfaces.
  • Double ended
  • PTFE coating
Specifications
Description Double Ended
Coating PTFE
Length (Metric) Blade 50 mm
Width (Metric) 4 mm
Width (Metric) Blade 2 mm
Diameter (Metric) Outer 2 mm
Length (Metric) 185 mm