Learn More
BRAND™ PTFE Coated Steel Core Stirring Rods
Product Code. p-8004613 Shop All Brand Products
Change view
Click to view available options
Length (Metric):
200 mm
250 mm
300 mm
400 mm
Unit Size:
Each
Product Code. Length (Metric) unitSize
10029880 300 mm Each
10363601 200 mm Each
10161871 250 mm Each
10272191 400 mm Each
4 options
Product Code. 10029880
Brand: BRAND™ 136325
Only null left
Add to basket
Add to basket
Unique high-density PTFE coated polyethylene stirring rods with a steel core with rounds ends.
  • PTFE coated steel core stirring rods with round end
  • High-density polyethylene (with steel core)
  • Can be bent as required
  • The shape is permanently retained
TRUSTED_SUSTAINABILITY
Specifications
Description PTFE Coated Steel Core
Diameter (Metric) 6 mm
Length (Metric) 300 mm
Quantity 1