Learn More
Bohlender™ PTFE Boiling Stones
2369.10 NOK
Specifications
Material PTFE
Quantity 500 g
View More Specs
Products 2
Product Code Brand Diameter (Metric) Price Quantity & Availability  
Product Code Brand Diameter (Metric) Price Quantity & Availability  
10194441
View Documents Promotion Details
Bohlender™
H972-02
4 mm
2369.10 NOK
Each
View alternative products
Save Up to 
»
Estimated Shipment: 07-06-2024
to see stock.
Add to basket
10524022
View Documents Promotion Details
Bohlender™
H972-04
6 mm
2369.10 NOK
Each
View alternative products
Save Up to 
»
Specifications
PTFE
500 g