Learn More
Bohlender™ PTFE Blunt Tip Tweezers
Click to view available options
Length (Metric):
150 mm
200 mm
Unit Size:
Each
Description
- Precast tweezers
- PTFE construction
- With pointed or blunt ends
- Universal chemical resistance
Specifications
Specifications
| Tip Style | Blunt |
| No. per Pack | Each |
| Design | Precast tweezer |
| Material | PTFE |
| Length (Metric) | 150 mm |
| Product Type | Tweezers |