Learn More
PTFE Beaker;Cap (ml), 250;Dia (mm), 66;Ht (mm), 97
Product Code. 15461477
Change view
Click to view available options
For Use With (Equipment):
1000mL beaker
100mL beaker
10mL beaker
2000mL beaker
250mL beaker
25mL beaker
500mL beaker
50mL beaker
Unit Size:
Each
Product Code. For Use With (Equipment) unitSize
15461477 10mL beaker Each
15471477 25mL beaker Each
15481477 50mL beaker Each
15491477 100mL beaker Each
15401487 250mL beaker Each
15411487 500mL beaker Each
15406175 1000mL beaker Each
15416175 2000mL beaker Each
8 options
Product Code. 15461477
Brand: Thermo Scientific Alfa Aesar 038084.KT
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use with beakers
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Available in a range of diameters
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 24mm
Material PTFE
For Use With (Equipment) 10mL beaker
Height (Metric) 33mm