Learn More
PTFE Beaker;Cap (ml), 250;Dia (mm), 66;Ht (mm), 97
Product Code. 15411487
Click to view available options
For Use With (Equipment):
1000mL beaker
100mL beaker
10mL beaker
2000mL beaker
250mL beaker
25mL beaker
500mL beaker
50mL beaker
Unit Size:
Each
Product Code. 15411487
Brand: Thermo Scientific Alfa Aesar 038089.KT
Request Bulk or Custom Format
For use with beakers
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Available in a range of diameters
TRUSTED_SUSTAINABILITY
Specifications
Diameter (Metric) Outer 80mm
Material PTFE
For Use With (Equipment) 500mL beaker
Height (Metric) 125mm