Learn More
PTFE Beaker Cover-Watch Glass;Dia (mm), 80;Fits Beakers, 250ml
Product Code. 15426175
Change view
Click to view available options
Description:
Watch glass; Fits 1000mL Beakers
Watch glass; Fits 100mL Beakers
Watch glass; Fits 2000mL Beakers
Watch glass; Fits 250mL Beakers
Watch glass; Fits 25mL Beakers
Watch glass; Fits 5-10mL Beakers
Watch glass; Fits 500mL Beakers
Watch glass; Fits 50mL Beakers
Unit Size:
Each
Product Code. Description unitSize
15426175 Watch glass; Fits 5-10mL Beakers Each
15496175 Watch glass; Fits 2000mL Beakers Each
15476175 Watch glass; Fits 500mL Beakers Each
15486175 Watch glass; Fits 1000mL Beakers Each
15436175 Watch glass; Fits 25mL Beakers Each
15446175 Watch glass; Fits 50mL Beakers Each
15456175 Watch glass; Fits 100mL Beakers Each
15466175 Watch glass; Fits 250mL Beakers Each
8 options
Product Code. 15426175
Brand: Thermo Scientific Alfa Aesar 038092.KT
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
For use as a beaker cover or a watch glass
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
  • Available in a range of diameters
TRUSTED_SUSTAINABILITY
Specifications
Description Watch glass; Fits 5-10mL Beakers
Material PTFE
Type Beaker