Learn More
Poulten & Graf GmbH PTFE Adaptor for FORTUNA™ Dispenser with Thread
Click to view available options
Thread Size:
GL 25
GL 28
GL 30
GL 33
GL 36
GL 52
GL 55
GL 60
GL 65
Unit Size:
Each
Description
- Suitable for OPTIFIX and POLYFIX dispenser
- Made of PTFE (Teflon™)
- Adapter with threads
- Can be used for acids and alkaline solutions that do not attack polypropylene
Specifications
Specifications
| Product Type | Adapters for OPTIFIX and POLYFIX Dispensers |
| Autoclavable | Autoclavable |
| Color | White |
| Description | Adapters for OPTIFIX and POLYFIX dispensers |
| Material | PTFE |
| For Use With (Equipment) | OPTIFIX and POLYFIX dispensers |
| Quantity | 1 |
| Thread Size | GL 60 |