missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PTER Monoclonal Antibody (OTI2F9), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813177
This item is not returnable.
View return policy
Description
PTER Monoclonal antibody specifically detects PTER in Human, Mouse, Rat samples. It is validated for Western BlotSpecifications
| PTER | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| Q60866, Q63530, Q96BW5 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 19212, 63852, 9317 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| OTI2F9 | |
| Unconjugated | |
| PTER | |
| AI790318; HPHRP; Mpr56-1; Parathion hydrolase-related protein; phosphotriesterase related; phosphotriesterase-related protein; Pter; resiniferatoxin-binding phosphotriesterase-related protein; resiniferatoxin-binding, phosphotriesterase-related; resiniferatoxin-binding, phosphotriesterase-related protein; resiniferotoxin-binding phosphotriesterase-related protein; Rpr1; Rpr-1 | |
| Human recombinant protein fragment corresponding to amino acids 1-242 of human PTER (NP_001001484) produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG1 |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction