missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PTAFR Polyclonal Antibody
GREENER_CHOICE

Product Code. 17170534 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μg
Unit Size:
100µg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17170534 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17170534 Supplier Invitrogen™ Supplier No. PA5112676

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

PTAFR Polyclonal Antibody for Western Blot, ICC/IF, IHC (P), ELISA

PTAFR shows structural characteristics of the rhodopsin gene family and binds platelet-activating factor (PAF). PAF is a phospholipid (1-0-alkyl-2-acetyl-sn-glycero-3-phosphorylcholine) that has been implicated as a mediator in diverse pathologic processes, such as allergy, asthma, septic shock, arterial thrombosis, and inflammatory processes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen PTAFR
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Gene PTAFR
Gene Accession No. P25105, Q62035
Gene Alias PAF receptor; PAFR; PAF-R; platelet activating factor receptor; platelet-activating factor receptor; Ptafr
Gene Symbols PTAFR
Host Species Rabbit
Immunogen Recombinant Human Platelet-activating factor receptor protein (297-342AA).
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19204, 5724
Target Species Human, Mouse
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.