Learn More
Testo™ Pt100 Robust Air Probe
Click to view available options
Temperature Range (Metric):
-50°C to +400°C
Unit Size:
Each
Specifications
Specifications
| Type | Probe, Robust Air |
| Certifications/Compliance | 60751 Standard |
| Response Time | 70 sec. |
| Temperature Range (Metric) | -50°C to +400°C |