Learn More
Psoralen, TRC
Product Code. 30135569
Change view
Click to view available options
Quantity:
10 mg
25 mg
50 mg
Unit Size:
10mg
25mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30135569 50 mg 50mg
30138279 25 mg 25mg
17942145 10 mg 10mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30135569 Supplier Toronto Research Chemicals Supplier No. TRCP83980050MG
 Restricted Item
Estimated Shipment: 05-08-2026
Add to basket
Add to basket
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 66-97-7
Molecular Formula C11 H6 O3
Molecular Weight (g/mol) 186.16
Synonym 7H-Furo[3,2-g][1]benzopyran-7-one, Furocoumarin (6CI), 2-Propenoic acid, 3-(6-hydroxy-5-benzofuranyl)-, δ-lactone, 6,7-Furanocoumarin, Ficusin, Furo[2',3':7,6]coumarin, Furo[4',5':6,7]coumarin, NSC 404562, Prosuler, Psoralen, Psoralene
IUPAC Name furo[3,2-g]chromen-7-one
SMILES O=C1Oc2cc3occc3cc2C=C1
Specifications
Quantity 50 mg
InChI Formula InChI=1S/C11H6O3/c12-11-2-1-7-5-8-3-4-13-9(8)6-10(7)14-11/h1-6H
Formula Weight 186.0317
Percent Purity >95%
Chemical Name or Material Psoralen
Grade HPLC