Learn More
PSMD4 Mouse anti-Human, CoraLite 594, Clone: 3G1A1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL594-66179-100UL
Product Code. 16870976
  • 5307.92 NOK / 100µL
Description
The PSMD4 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human PSMD4. This antibody recognizes human antigen. The PSMD4 antibody has been validated for the following applications: IF analysis. This antibody is coralite™594.
Specifications
Specifications
PSMD4
Monoclonal
2000 μg/mL
Mouse
Primary
Human
Immunofluorescence
3G1A1
CoraLite 594
100 μL
5710
IgG1