missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSMC6 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15839-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
PSMC6 (proteasome (prosome, macropain) 26S subunit, ATPase 6), also known as P44, p42, SUG2, S10B or CADP44, is a regulatory component of the 26S proteasome. More specifically, PSMC6 is one of the six ATPase subunits of the 19S regulator base. It contains a leucine zipper motif and an AAA (ATPase associated with diverse cellular activities) domain and belongs to the AAA ATPase family of chaperone-like ATPases.Specifications
PSMC6 | |
Polyclonal | |
Unconjugated | |
PSMC6 | |
CADP44, p42, P44, Proteasome subunit p42, PSMC6, Rpt4, SUG2 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
289990, 5706, 67089 | |
-20°C | |
Liquid |
Immunocytochemistry, Immunofluorescence, Western Blot | |
0.21 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P62333, P62334 | |
Psmc6 | |
PSMC6 Fusion Protein Ag8589 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |