missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ PSMC4 (Human) Recombinant Protein
Human PSMC4 full-length ORF ( AAH00343, 1 a.a. - 418 a.a.) recombinant protein with GST-tag at N-terminal.
Brand: Abnova™ H00005704-P01.10ug
Additional Details : Weight : 0.00010kg
Description
- Sequence: MEEIGILVEKAQDEIPALSVSRPQTGLSFLGPEPEDLEDLYSRYKKLQQELEFLEVQEEYIKDEQKNLKKEFLHAQEEVKRIQSIPLVIGQFLEAVDQNTAIVGSTTGSNYYVRILSTIDRELLKPNASVALHKHSNALVDVLPPEADSSIMMLTSDQKPDVMYADIGGMDIQKQEVREAVELPLTHFELYKQIGIDPPRGVLMYGPPGCGKTMLAKAVAHHTTAAFIRVVGSEFVQKYLGEGPRMVRDVFRLAKENAPAIIFIDEIDAIATKRFDAQTGADREVQRILLELLNQMDGFDQNVNVKVIMATNRADTLDPALLRPGRLDRKIEFPLPDRRQKRLIFSTITSKMNLSEEVDLEDYVARPDKISGADINSICQESGMLAVRENRYIVLAKDFEKAYKTVIKKDEQEHEFYK
Specifications
AAH00343 | |
proteasome (prosome, macropain) 26S subunit, ATPase, 4 | |
125% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PSMC4 | |
GST |
5704 | |
Wheat germ expression system | |
10 μg | |
MGC13687, MGC23214, MGC8570, MIP224, S6, TBP7 | |
Wheat Germ (in vitro) | |
50 mM Tris-HCI, 10 mM reduced Glutathione, pH 8.0 in the elution buffer |