missing translation for 'onlineSavingsMsg'
Learn More

PSG1 Rabbit anti-Human, Polyclonal, Proteintech

Product Code. p-7204274 Shop All Proteintech Products
Change view
Click to view available options
Quantity:
150 μL
20 μL
Unit Size:
150µL
20µL
Product Code. Quantity unitSize
16839133 20 μL 20µL
16829133 150 μL 150µL
2 options
This item is not returnable. View return policy

Product Code. 16839133

Brand: Proteintech 118091AP20UL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Human pregcy-specific glycoproteins (PSG), are a group of closely related secreted glycoproteins which are highly expressed in placental syncytiotrophoblast cells of fetal origin. PSG are commonly expressed in tumors of trophoblast origin (hydatidiform mole, choriocarcinoma). They represent the most abundant fetal protein in the maternal blood at term. The eleven human PSG genes (PSG1-PSG11) form together with the carcinoembryonic antigen (CEA)-related cell adhesion molecule (CEACAM) genes the CEACAM gene family. PSG stimulates secretion of TH2-type cytokines from monocytes. CD9 was shown to represent the monocyte receptor for murine PSG17. PSG are thought to modulate the maternal immune system during pregcy thus protecting the semiallotypic fetus from rejection.

Specifications

Antigen PSG1
Applications Immunocytochemistry, Immunofluorescence, Western Blot, Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.13 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
Gene PSG1
Gene Accession No. P11464
Gene Alias B1G1, CD66f, DHFRP2, FL NCA 1/2, PBG1, PS beta C/D, PS beta G 1, PSBG 1, PSBG1, PSG1, PSG95, PSGGA, PSGIIA, SP1
Gene Symbols PSG1
Host Species Rabbit
Immunogen PSG1 Fusion Protein Ag2468
Purification Method Antigen Affinity Chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5669
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.