missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PSAT1 Rabbit anti-Human, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 10501-1-AP-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is likely a phosphoserine aminotransferase, based on similarity to proteins in mouse, rabbit, and Drosophila. Alternative splicing of this gene results in two transcript variants encoding different isoforms.Specifications
| PSAT1 | |
| Polyclonal | |
| Unconjugated | |
| PSAT1 | |
| EPIP, Phosphoserine aminotransferase, PSA, PSAT, PSAT1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 293820, 29968 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Immunofluorescence, Western Blot, Immunocytochemistry, Immunohistochemistry (Paraffin) | |
| 0.33 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q9Y617 | |
| Psat1 | |
| PSAT1 Fusion Protein Ag0776 | |
| 20 μL | |
| Primary | |
| Rat, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction