Learn More
PS-1145, MedChemExpress
Product Code. 16483467
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
25 mg
50 mg
100 mg
Unit Size:
100mg
10mg
25mg
50mg
5mg
Product Code. 16483467
Brand: Medchem Express HY18008
MedChemExpress PS-1145 is an IκB kinase (IKK) inhibitor with an IC50 of 88 nM.
PS-1145 is an IκB kinase (IKK) inhibitor with an IC50 of 88 nM.
Specifications
Chemical Name or Material PS-1145
CAS 431898-65-6
Color Light Yellow
Purity Grade Notes Research
Molecular Formula C17H11ClN4O
Quantity 50 mg
Solubility Information DMSO : 20.83 mg/mL (64.54 mM; Need ultrasonic)
SMILES O=C(C1=CC=CN=C1)NC2=CC(Cl)=CC3=C2NC4=C3C=CN=C4
Molecular Weight (g/mol) 322.75
Formula Weight 322.75
Percent Purity 99.63%
Grade Research
Physical Form Powder
For Use With (Application) Cancer-Kinase/protease
Show More Show Less
Research purposes only
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.