missing translation for 'onlineSavingsMsg'
Learn More

PRSS8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 15527-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16884734

  • 3468.15 NOK / 150µL
Estimated Shipment: 14-12-2022
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

This antibody can recognize 39 kDa monomer form and 70 kDa homodimer form.

This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.
Specifications

Specifications

PRSS8
Polyclonal
Unconjugated
PRSS8
CAP1, PROSTASIN, protease, serine, 8, PRSS8, Serine protease 8
Rabbit
Antigen Affinity Chromatography
RUO
192107, 5652, 76560
-20°C
Liquid
Western Blot, Immunohistochemistry (Paraffin)
0.35 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Q16651, Q9ES87, Q9ESD1
PRSS8
PRSS8 Fusion Protein Ag7234
150 μL
Primary
Rat, Mouse, Human
Antibody
IgG
Videos
SDS
Documents
Certificates
Special Offers

Special Offers