missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRSS8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15527-1-AP-20UL
Additional Details : Weight : 0.00010kg
Description
This antibody can recognize 39 kDa monomer form and 70 kDa homodimer form.
This gene encodes a trypsinogen, which is a member of the trypsin family of serine proteases. This enzyme is highly expressed in prostate epithelia and is one of several proteolytic enzymes found in seminal fluid. The proprotein is cleaved to produce a light chain and a heavy chain which are associated by a disulfide bond. It is active on peptide linkages involving the carboxyl group of lysine or arginine.Specifications
PRSS8 | |
Polyclonal | |
Unconjugated | |
PRSS8 | |
CAP1, PROSTASIN, protease, serine, 8, PRSS8, Serine protease 8 | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
192107, 5652, 76560 | |
-20°C | |
Liquid |
Western Blot, Immunohistochemistry (Paraffin) | |
0.35 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
Q16651, Q9ES87, Q9ESD1 | |
PRSS8 | |
PRSS8 Fusion Protein Ag7234 | |
20 μL | |
Primary | |
Human, Rat, Mouse | |
Antibody | |
IgG |