missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ PRSS23 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15704431 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
Product Code. Quantity unitSize
15704431 100 μL 100µL
1 options
This item is not returnable. View return policy

Product Code. 15704431

Brand: Invitrogen™ PA556516

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: PWKPTWPAYR LPVVLPQSTL NLAKPDFGAE AKLEVSSSCG PQCHKGTPLP TYEEAKQYLS YETLYANGSR TETQVGIYIL SSSGDGA Highest antigen sequence identity to the following orthologs: Mouse - 91%, Rat - 91%.

TRUSTED_SUSTAINABILITY

Specifications

Antigen PRSS23
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.10 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene PRSS23
Gene Accession No. O95084
Gene Alias protease, serine 23; protease, serine, 23; PRSS23; Putative secreted protein Zsig13; serine protease 23; serine protease, umbilical endothelium; SIG13; SPUVE; UNQ270/PRO307; ZSIG13
Gene Symbols PRSS23
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human PRSS23. Recombinant protein control fragment (Product #RP-95860).
Purification Method Antigen Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11098
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.