missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRSS2 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 15005-1-AP-150UL
Additional Details : Weight : 0.00010kg
Description
This gene belongs to the trypsin family of serine proteases and encodes anionic trypsinogen. It is part of a cluster of trypsinogen genes that are located within the T cell receptor beta locus. Enzymes of this family cleave peptide bonds that follow lysine or arginine residues. This protein is found at high levels in pancreatic juice and its upregulation is a characteristic feature of pancreatitis. This protein has also been found to activate pro-urokinase in ovarian tumors, suggesting a function in tumor invasion. In addition, this enzyme is able to cleave across the type II collagen triple helix in rheumatoid arthritis synovitis tissue, potentially participating in the degradation of type II collagen-rich cartilage matrix. Alternative splicing results in multiple transcript variants.Specifications
PRSS2 | |
Polyclonal | |
Unconjugated | |
PRSS2 | |
Anionic trypsinogen, PRSS2, Serine protease 2, TRY2, TRY8, TRYP2, Trypsin 2, Trypsin II | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
22072, 25052, 5645 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.27 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
P00763, P07146, P07478 | |
PRSS2 | |
PRSS2 Fusion Protein Ag6461 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |