missing translation for 'onlineSavingsMsg'
Learn More

PRPS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 27024-1-AP-150UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16895325

  • 3828.53 NOK / 150µL
Estimated Shipment: 10-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

The function remains unknown.
Specifications

Specifications

PRPS2
Polyclonal
Unconjugated
PRPS2
PPRibP, PRPS2, PRS II, PRSII
Rabbit
Antigen Affinity Chromatography
RUO
110639, 5634
-20°C
Liquid
Western Blot
0.27 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
P11908, Q9CS42
PRPS2
PRPS2 Fusion Protein Ag25785
150 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers