missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPS2 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 27024-1-AP-20UL
This item is not returnable.
View return policy
Description
The function remains unknown.Specifications
| PRPS2 | |
| Polyclonal | |
| Unconjugated | |
| PRPS2 | |
| PPRibP, PRPS2, PRS II, PRSII | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 110639, 5634 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.27 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| P11908, Q9CS42 | |
| PRPS2 | |
| PRPS2 Fusion Protein Ag25785 | |
| 20 μL | |
| Primary | |
| Mouse, Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction