missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPS1L1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16657-1-AP-20UL
This item is not returnable.
View return policy
Description
Catalyzes the synthesis of phosphoribosylpyrophosphate (PRPP) that is essential for nucleotide synthesis.Specifications
| PRPS1L1 | |
| Polyclonal | |
| Unconjugated | |
| PRPS1L1 | |
| PRPS1, PRPS1 like 1, PRPS1L1, PRPS3, PRPSL, PRS III | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 221823, 298954, 75456 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 0.17 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P21108 | |
| Prps1l1 | |
| PRPS1L1 Fusion Protein Ag10111 | |
| 20 μL | |
| Primary | |
| Rat, Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction