missing translation for 'onlineSavingsMsg'
Learn More
Learn More
PRPF40A Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 17392-1-AP-150UL
This item is not returnable.
View return policy
Description
FNBP3, Formin-binding protein 3, is a 957 amino acid protein encoded by the human gene PRPF40A. FNBP3 belongs to the PRPF40 family and contains five FF domains and two WW domains. Nuclear proteins harboring both WW and FF protein interaction modules bind to splicing factors as well as RNA polymerase II and may serve to link transcription with splicing. Through the WW domains, FNBP3 will bind to the Formin proline-rich regions. Also known as Pre-mRNA-processing factor 40 homolog A, FNBP3 binds to WASL/N-WASP (Neuronal Wiskott-Aldrich syndrome protein) complex and suppresses its translocation from the nucleus to the cytoplasm, thereby inhibiting its cytoplasmic function. FNBP3 is widely expressed in most tissues and is localized to the nuclear speckles.Specifications
| PRPF40A | |
| Polyclonal | |
| Unconjugated | |
| PRPF40A | |
| Fas ligand associated factor 1, FBP 11, FBP11, FLAF1, FNBP3, Formin binding protein 11, Formin binding protein 3, HIP 10, HIP10, Huntingtin yeast partner A, HYPA, NY REN 6, PRPF40A | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 55660, 56194 | |
| -20°C | |
| Liquid |
| Immunoprecipitation, Western Blot | |
| 0.2 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| O75400, Q9R1C7 | |
| PRPF40A | |
| PRPF40A Fusion Protein Ag11290 | |
| 150 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction