missing translation for 'onlineSavingsMsg'
Learn More

PRPF40A Rabbit anti-Human, Mouse, Polyclonal, Proteintech

Rabbit Polyclonal Antibody

Brand:  Proteintech 17392-1-AP-20UL

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16847274

  • 1415.13 NOK / 20µL
Estimated Shipment: 16-10-2023
to see stock.

Please to purchase this item. Need a web account? Register with us today!


Description

Description

FNBP3, Formin-binding protein 3, is a 957 amino acid protein encoded by the human gene PRPF40A. FNBP3 belongs to the PRPF40 family and contains five FF domains and two WW domains. Nuclear proteins harboring both WW and FF protein interaction modules bind to splicing factors as well as RNA polymerase II and may serve to link transcription with splicing. Through the WW domains, FNBP3 will bind to the Formin proline-rich regions. Also known as Pre-mRNA-processing factor 40 homolog A, FNBP3 binds to WASL/N-WASP (Neuronal Wiskott-Aldrich syndrome protein) complex and suppresses its translocation from the nucleus to the cytoplasm, thereby inhibiting its cytoplasmic function. FNBP3 is widely expressed in most tissues and is localized to the nuclear speckles.
Specifications

Specifications

PRPF40A
Polyclonal
Unconjugated
PRPF40A
Fas ligand associated factor 1, FBP 11, FBP11, FLAF1, FNBP3, Formin binding protein 11, Formin binding protein 3, HIP 10, HIP10, Huntingtin yeast partner A, HYPA, NY REN 6, PRPF40A
Rabbit
Antigen Affinity Chromatography
RUO
55660, 56194
-20°C
Liquid
Immunoprecipitation, Western Blot
0.2 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
O75400, Q9R1C7
PRPF40A
PRPF40A Fusion Protein Ag11290
20 μL
Primary
Human, Mouse
Antibody
IgG
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers