Learn More
Hamilton™ PRP-X200 10 μm 2.1 x 150 mm
Click to view available options
Length (Metric):
150 mm
Unit Size:
Each
Description
- Separation Mode-Cation Exchange
- Stationary Phase-PRP-X200
- Format-HPLC Column
- Hardware Inner Diameter-2.1 mm
- Hardware Length-150 mm
- Particle Size-10 μm
- USP Classification-USP L22
- Packing Material Type-PSDVB/Sulfonic Acid
- Pore Size-100 Å
- Hardware Material-Stainless Steel
- Max Pressure-5,000 psi
- Exchange Capacity-35 μeq/gm
- Mobile Phase Limits-pH 1-13. 0-100% aqueous, organic modifier
- Buffer Strength-0.0-0.5N
- Temp Limits-5-60°C
- Restoration Inject several times with 100 μL of 1 N nitric acid
- Compound Classification-Organic/Inorganic Cations
- UOM-1 EACH
Specifications
Specifications
| Column Format | HPLC Column |
| Diameter (Metric) | 2.1 mm |
| Length (Metric) | 150 mm |
| Max. Pressure | 5,000 psi |
| Particle Size | 10 μm |
| pH | pH 1-13. 0-100% aqueous, organic modifier |
| Pore Size | 10 Å |
| Temperature | 5° to 60°C |
| USP Type | USP L22 |
| Packing Material | PSDVB/Sulfonic Acid |
| Show More |