missing translation for 'onlineSavingsMsg'
Learn More

Hamilton™ PRP-1 10 μm 100 Å 10 x 100 mm

Product Code. 11538084
Change view
Click to view available options
Length (Metric):
100 mm
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Length (Metric) Unit Size
11538084 100 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 11538084 Supplier Hamilton™ Supplier No. 79499
Estimated Shipment: 22-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

PRP-1 columns have inherent reversed-phase separation qualities that eliminate the requirement for a stationary phase coating. Hydrolysis of the stationary phase coating is often the limiting factor in column lifetime, so the lack of coating on the PRP-1 resin extends the longevity of the column. PRP-1 also have exceptional sample recovery (≥ 95%).

Specification Sheet

  • Separation Mode – Reversed-Phase
  • Stationary Phase – PRP-1
  • Format – HPLC Column
  • Hardware Inner Diameter – 10 mm
  • Hardware Length – 100 mm
  • Particle Size – 10 μm
  • USP Classification – USP L21
  • Packing Material Type – PSDVB
  • Pore Size – 100 Å
  • Hardware Material – Stainless Steel
  • Max Pressure – 5,000 psi
  • Mobile Phase Limits – pH 1-13. 0-100% aqueous, organic modifier
  • Buffer Strength – 0.0-0.5N
  • Temp Limits – 5-85°C
  • Restoration – Run a gradient of 100% water to 100% acetonitrile. Repeat three times
  • Compound Classification – Organics
  • UOM – 1 EACH

Specifikationer

Column Format HPLC Column
Diameter (Metric) 10 mm
Length (Metric) 100 mm
Max. Pressure 5,000 psi
Particle Size 10 μm
pH pH 1-13. 0-100% aqueous, organic modifier
Pore Size 100 Å
Temperature 5° to 85°C
USP Type USP L21
Packing Material PSDVB
Phase Reversed Phase
Material Stainless Steel
Product Type PRP-1 Columns
Visa mer Visa mindre
Produkttitel
Välj ett problem

Genom att klicka på Skicka bekräftar du att du kan bli kontaktad av Fisher Scientific angående feedbacken du har lämnat i detta formulär. Vi kommer inte att dela din information för andra ändamål. All kontaktinformation som tillhandahålls ska också underhållas i enlighet med vår Sekretesspolicy.