Learn More
Invitrogen™ RNase-free Tips, 1000 μL size
Click to view available options
Quantity:
10 racks
Unit Size:
Each
Description
- Ambion Tips are RNase- and DNase-free and are provided in ten racks containing 96 tips each
- They are compatible with Eppendorf, Finnpipette, Oxford Benchmate, Pipetman, Proline, Rainin, Socorex, and other brands of pipettors
- Each lot of tips is subjected to rigorous testing for RNase contamination and is certified to be nuclease-free
Specifications
Specifications
| Volume (Metric) | 1000 μL |
| Filtered | Non-filtered |
| Sterility | Sterile |
| Quantity | 10 racks |
| Product Type | RNase-free Tips |
| Content And Storage | Includes 10 racks with 96 tips each. Store at room temperature. |
| Shipping Condition | Room Temperature |
| For Use With (Equipment) | Universal Fit |
| Product Line | Ambion |